Sodium iodide, ultra dry ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

013604-06 013604-14 013604-22
AA13604-06EA 88.4 USD
AA13604-06 AA13604-14 AA13604-22
Sodium iodide, ultra dry ≥99.999% (metals basis)
Sodium iodide

Purity: 99.999% CAS No.: 7681-82-5 Molecular Formula: NaI Formula Weight: 149.89 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.67 Refractive Index: 1.7745 Sensitivity: Light Sensitive and Hygroscopic Appearance: White Odor: Odorless Melting Point: 651° Boiling Point: 1300-1304° Merck Reference: 14,8631 MDL No.: MFCD00003532

Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1300 °C (1013 hPa)
Melting Pt: 662 °C
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 13,08705

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR