Indium ≥99.9999% (metals basis), shot, Puratronic® 4-8 mm (0.2-0.3 in)
Supplier: Thermo Scientific Chemicals
011359-KN
011359-KM
AA11359-KNPK
1176
USD
AA11359-KN
AA-11359-KM
AA11359-KM
Indium ≥99.9999% (metals basis), shot, Puratronic® 4-8 mm (0.2-0.3 in)
Indium
Purity: 99.9999%
CAS No.: 7440-74-6
Odor: Odorless
MDL No.: MFCD00134048
Formula:
In MW: 114.82 g/mol Boiling Pt: 2000 Melting Pt: 156 Density: 7.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...