Gastight® 1700 Series Syringe Replacement Plungers, Hamilton Company
Supplier: Hamilton
13205 1162-01 1162-02 1162-03 1117-03 1122-01 1169-01 1117-02 13269 1117-01
60373-195EA 46.59 USD
60373-195 60373-638 60373-642 60373-646 60373-648 60373-667 60373-650 89221-372 89221-376 89221-370
Gastight® 1700 Series Syringe Replacement Plungers, Hamilton Company
Syringe Barrels, Plungers and Pistons
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
These replacement plungers are for Hamilton 1700 series Gastight® syringes ranging in volume from 10 to 500 µl.
Each is manufactured to achieve the highest level of accuracy and precision possible.