Nunc® Cryobank Vials, Thermo Scientific
Supplier: Thermo Fisher Scientific
374220 374222 374223 374221LOCK
89134-760CS 1254.35 USD
89134-760 89134-764 89134-766 89218-754
Nunc® Cryobank Vials, Thermo Scientific
Vials
Thermo Scientific Nunc® Cryobank vial systems are automation-friendly.
The code will work with any Thermo Scientific 2D reader and allows for safe sample identification and reading of whole racks in one process. The tubes click into the rack and can be handled without the risk of accidentally dropping a vial. They have the same dimensions as traditional Nunc® Cryovials and can be stored in existing boxes and racks.
They provide the customer with superior tracking of their cryovials using a 2D code, the highest storage density on the market and the ability to automate the capping and decapping of 48 vials in one operation with the Thermo Scientific 48 way Capit-All decapper.
Certificaciones: CE marked for diagnostic use; certified DNAse, RNAse free.
Información para pedidos: Vials come racked.