Nalgene® Narrow Mouth Bottles in Shrink-Wrap Trays, Natural HDPE, Thermo Scientific
Supplier: Thermo Fisher Scientific
322089-0001 322089-0002 322089-0008
16059-632CS 912.96 USD
16059-632 16059-634 16059-638
Nalgene® Narrow Mouth Bottles in Shrink-Wrap Trays, Natural HDPE, Thermo Scientific
Bottles Plastic Bottles
Nalgene® high density polyethylene narrow-mouth bottles in shrink-wrap trays are leak proof.
They are packaged in rigid trays. The bottles contain natural polypropylene closures and are ready to feed into filling lines for various reagents.
Certificaciones: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Información para pedidos: They are bulk packed for packaging applications.