ABTS Microwell Peroxidase Substrate Kit, KPL
Supplier: KPL
5120-0033 5150-0017 5120-0032 5120-0041 5120-0043 5120-0046
95059-130EA 1285.03 USD
95059-130 95059-196 95059-128 95059-142 95059-144 95059-146
ABTS Microwell Peroxidase Substrate Kit, KPL
Assays ELISAs
A good, all-purpose substrate that offers the widest working range
ABTS Peroxidase Stop Solution contains 5% sodium dodecyl sulfate (SDS) in water. This solution is specifically designed for use with KPL’s ABTS Peroxidase Substrates (both 1- and 2-component) to stop development in ELISA plate wells.
Ideal when low background is required and moderate sensitivity is acceptable. ABTS develops a blue-green color in the presence of peroxidase labeled conjugates measurable between 405 and 410 nm. No change in color when reaction is stopped. Recommended for ELISA applications. Single component format that contains 2, 2'-azino-di(3-ethylbenzthiazoline-6-sulfonate), no reagent peparation required.