Nalgene® Packaging Bottles, Amber, Wide Mouth, Translucent, HDPE, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
312184-0004 312184-0008 312184-0016
16059-564CS 614.4 USD
16059-564 16059-566 16059-568
Nalgene® Packaging Bottles, Amber, Wide Mouth, Translucent, HDPE, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene® Amber wide-mouth packaging bottles are high density polyethylene.
The amber color protects light sensitive products, and the wide mouth allows for easy filling and dispensing of liquids and powders. The bottles come with opaque amber polypropylene closures.
Certificaciones: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Información para pedidos: They are bulk packed for packaging applications.