Genistein

Supplier: R&D Systems

1110/10 1110/50
100505-256EA 117.14 USD
100505-256 100509-080
Genistein
Genistein
Formula: C₁₅H₁₀O₅
Melting Pt: 297…298 °C
MDL Number: MFCD00016952
CAS Number: 446-72-0
EINECS: 207-174-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR