3-Bromo-7-nitro-1H-indazole

Supplier: R&D Systems

Synonyms: 3-Bromo-7-nitro-1H-indazole, 3-Bromo-7-nitroindazole

0735/10 0735/50
100506-818EA 263.2 USD
100506-818 100508-534
3-Bromo-7-nitro-1H-indazole
3-Bromo-7-nitro-1H-indazole
Formula: C₇H₄BrN₃O₂
MW: 242.03 g/mol
Melting Pt: 176 °C
MDL Number: MFCD00159910
CAS Number: 74209-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR