Zinc selenide, crystals for spectrophotometry
Supplier: INTERNATIONAL CRYSTAL LABS MS
|
Danger
|
Synonyms:
Zinc (II) selenide
0003J-4116
0003J-4115
0003J-4117
100213-248EA
1914.62
USD
100213-248
100213-246
100213-250
Zinc selenide, crystals for spectrophotometry
Zinc selenide
Zinc Selenide (ZnSe) is commonly used for CO2 laser focusing lenses, night vision applications, ATR prisms and transmission windows for IR spectroscopy.
Formula:
SeZn MW: 144.35 g/mol Melting Pt: 1793 Density: 5.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011296 CAS Number: 1315-09-9 EINECS: 215-259-7 UN: 3283 ADR: 6.1 III Merck Index: 12,10290 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...