Calcium fluoride, crystal rectangle for spectrophotometry, polished
Supplier: INTERNATIONAL CRYSTAL LABS MS
|
Warning
|
0002D-295
0002D-142
100212-724EA
258.4
USD
100212-724
100212-716
Calcium fluoride, crystal rectangle for spectrophotometry, polished
Calcium fluoride
International Crystal Labs celebrated its 40th anniversary in 2002. They have produced quality crystal optics for spectroscopy and CO2 lasers since 1962. ICL operates a vertically integrated crystal optics production plant and grow their own crystals.
Formula:
CaF₂ Boiling Pt: 2500 °C (1013 hPa) Melting Pt: 1418 °C Density: 3.18 g/cm³ (20 °C) |
MDL Number:
MFCD00010907 CAS Number: 7789-75-5 EINECS: 232-188-7 Merck Index: 13,01669 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...