Xanthine ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine
XA110-100GM
XA110-25GM
700008-840EA
481.6
USD
700008-840
700008-842
Xanthine ≥98%
Xanthine
Xanthine, also known as 1H-Purine-2,6-dione, is a purine base found in human body fluids and tissues.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₅H₄N₄O₂ MW: 152.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00078453 CAS Number: 69-89-6 EINECS: 200-718-6 Merck Index: 12,10193 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...