Uric acid ≥99%, Reagent Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1H-Purine-2,6,8(3H,7H,9H)-trione

UR120-1KG UR120-500GM UR120-100GM
700009-024EA 2109.6 USD
700009-024 700009-030 700009-022
Uric acid ≥99%, Reagent Grade for synthesis
Uric acid

Uric Acid, Reagent is a product of the metabolic breakdown of purine nucleotides. High blood concentrations of uric acid can lead to gout. The chemical is associated with other medical conditions including diabetes and the formation of ammonium acid urate kidney stones. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₅H₄N₄O₃
MW: 168.11 g/mol
Melting Pt: 436
Density: 1.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005712
CAS Number: 69-93-2
EINECS: 200-720-7
Merck Index: 12,10014

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR