TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether) for electrophoresis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

TR120-4LTPL TR120-500MLP TR120-20LTMT
700011-170EA 501.95 USD
700011-170 700011-172 700011-168
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether) for electrophoresis
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether)

Triton(R) X-114, Electrophoresis Grade – The Electrophoresis Grade designation signifies that this material is specifically suitable for use in electrophoresis applications.

Boiling Pt: ? 150 °C (1013 hPa)
Density: 1.055 g/cm³ (20 °C)
Flash Pt: ≥ 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00128254
CAS Number: 9036-19-5
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR