Titanium(IV) oxide ≥99% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide

TI140-12KG TI140-2.5KG TI140-45KGBL TI140-500GM
700000-872EA 1244.96 USD
700000-872 95036-328 89079-044 95036-330
Titanium(IV) oxide ≥99% USP
Titanium(IV) oxide

Titanium Dioxide, USP is used as a food coloring. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: TiO₂
MW: 79.88 g/mol
Boiling Pt: 2900 °C (1013 hPa)
Melting Pt: 1855 °C
Density: 4.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR