D-(+)-Tyrosine ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: H-D-Tyr-OH

T3180-1GM T3180-5GM
700007-498EA 97.05 USD
700007-498 700007-500
D-(+)-Tyrosine ≥98%
D-(+)-Tyrosine

D-Tyrosine Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00063073
CAS Number: 556-02-5
EINECS: 209-112-6
Merck Index: 12,09970

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR