D-α-Tocopherol
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
(+)-α-Tocopherol, Vitamin E Alcohol, (R)-2,5,7,8-Tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, α-Vitamin E, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E, D(+)-α-Tocopherol
(+)-alpha-Tocopherol is a type of vitamin E which is accumulated and absorbed in humans. Alpha-tocopherol has an E number, E307, and is used as an antioxidant.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₂₉H₅₀O₂ MW: 430.71 g/mol Density: 0.950 Storage Temperature: Freezer |
MDL Number:
MFCD00072045 CAS Number: 59-02-9 EINECS: 233-466-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...