2,2'-Thiodiacetic acid for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Thiodiglycolic acid, Thiodiacetic Acid

T2473-100GM T2473-25GM
700008-216EA 261.85 USD
700008-216 700008-218
2,2'-Thiodiacetic acid for synthesis
2,2'-Thiodiacetic acid

Thiodiacetic Acid is used as an analytical reagent for the detection of copper, lead, mercury, silver.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₆O₄S
MW: 150.16 g/mol
Boiling Pt: 168
Melting Pt: 128…131 °C
Density: 1.661 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004359
CAS Number: 123-93-3
EINECS: 204-663-9
UN: 3261
ADR: 8 III
Merck Index: 12,09467

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR