Triisopropanolamine ≥99%

Supplier: Spectrum Chemical Mfg. Corp.

T2120-1KG T2120-100GM T2120-200LTB T2120-20LT
700007-068EA 106.4 USD
700007-068 700007-066 700007-070 700007-072
Triisopropanolamine ≥99%
Triisopropanolamine

Triisopropanolamine, also known as 1-Amino-2-propanol, is an amino alcohol and can be used in several applications to achieve buffering, basicity, and alkalinity objectives.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₂₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD00004533
CAS Number: 122-20-3
EINECS: 204-528-4

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR