Thiosalicylic acid ≥97%

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 2-Thiobenzoic acid, 2-Mercaptobenzoic acid

T2014-100GM T2014-25GM T2014-500GM T2014-50KGBL
700008-198EA 313.45 USD
700008-198 700008-202 700008-204 700008-206
Thiosalicylic acid ≥97%
Thiosalicylic acid

Thiosalicylic Acid, also known as o-Thiosalicylic acid, is a precursor to the dyestuff thioindigo and the vaccine preservative thiomersal.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₆O₂S
MW: 154.19 g/mol
Boiling Pt: 230 °C (0.02 -rr)
Melting Pt: 165 - 166 °C
Density: 1.489 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004836
CAS Number: 147-93-3
EINECS: 205-704-3
UN: 3335
ADR: 9,
Merck Index: 12,09498

Order Now

Specification Test Results

Assay 97% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR