Trichloroacetic acid (TCA) 30% (w/v) in aqueous solution

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

T-181-500MLG T-181-1LTGL
700004-670EA 194.77 USD
700004-670 700004-668
Trichloroacetic acid (TCA) 30% (w/v) in aqueous solution
Trichloroacetic acid (TCA)

Trichloroacetic Acid, 30 Percent (w/v) Aqueous Solution - Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability

Formula: Cl₃CCOOH
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: Max. 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 2564
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=1bea41ad50581ecec5e4568389942c56b92e3f0f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR