1,2,4-Trihydroxybenzene ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Benzene-1,2,4-triol, 1,2,4-Benzenetriol

T1346-25GM T1346-5GM
700003-336EA 1322.85 USD
700003-336 700003-334 700003-338
1,2,4-Trihydroxybenzene ≥99%
1,2,4-Trihydroxybenzene

1,2,4-Trihydroxybenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. For cosmetic products, it is used as a hair dye.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 362 °C (1013 hPa)
Melting Pt: 140 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002198
CAS Number: 533-73-3
EINECS: 208-575-1
Merck Index: 13,01073

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR