1,3,3,5-Tetramethyl-1,1,5,5-tetraphenyltrisiloxane

Supplier: SPECTRUM CHEMICAL MFG CORP

T1293-25GM T1293-100GM T1293-5GM
700015-916EA 148.51 USD
700015-916 700015-914 700015-918
1,3,3,5-Tetramethyl-1,1,5,5-tetraphenyltrisiloxane
1,3,3,5-Tetramethyl-1,1,5,5-tetraphenyltrisiloxane

1,3,3,5-Tetramethyl-1,1,5,5-tetraphenyltrisiloxane is used as an emollient in cosmetics and as a diffusion pump fluid.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₈H₃₂O₂Si₃
Density: 1.070
Storage Temperature: Ambient
MDL Number: MFCD00053661
CAS Number: 3982-82-9
EINECS: 223-620-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=457cd781e6a3faed6aad60c86c29c9f7f6244861&vid=457cd781e6a3faed6aad60c86c29c9f7f6244861&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR