Triphenyl phosphate, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.
Warning

T1121-250GM T1121-5KG T1121-1KG
700006-670EA 124.2 USD
700006-670 700006-672 700006-668
Triphenyl phosphate, Reagent Grade
Triphenyl phosphate
Formula: C₁₈H₁₅O₄P
Boiling Pt: 412 °C (1013 hPa)
Melting Pt: 50…51 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 223 °C
Storage Temperature: Ambient
MDL Number: MFCD00003031
CAS Number: 115-86-6
EINECS: 204-112-2
UN: 3077
ADR: 9,III
Merck Index: 12,09872

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR