1-Octadecanol ≥90% NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol

ST115-500GM ST115-2.5KG ST115-12KG
95036-082EA 142.88 USD
95036-082 95036-080 700002-728
1-Octadecanol ≥90% NF
1-Octadecanol

BSE/TSE statement (available upon request) Allergen statement (available upon request)


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: H₃C(CH₂)₁₆CH₂OH
MW: 270.5 g/mol
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 56 - 57 °C
Density: 0.805 to 0.815 g/cm³ (60 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002823
CAS Number: 112-92-5
EINECS: 204-017-6
Merck Index: 12,08960

Order Now

Specification Test Results

Assay 90% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR