Stearic acid, powder NF Type 50
Supplier: Spectrum Chemical Mfg. Corp.
Stearic Acid, Type 50, Powder, NF is used as an emulsifying agent, solubilizing agent, tablet and capsule lubricant. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₁₈H₃₆O₂ Boiling Pt: 361 °C (1013 hPa) Melting Pt: 67 °C Density: 0.94 g/cm³ (20 °C) Flash Pt: 196 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00002752 CAS Number: 57-11-4 EINECS: 200-313-4 Merck Index: 13,08882 |
|
Specification Test Results
Assay | 40% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...