Sorbitan monostearate NF

Supplier: Spectrum Chemical Mfg. Corp.

SPA63-12KG SPA63-2.5KG SPA63-500GM SPA63-90KGBL
700009-736EA 1771.1 USD
700009-736 95036-020 95036-022 76014-918
Sorbitan monostearate NF
Sorbitan monostearate

BSE/TSE statement (available upon request) Allergen statement (available upon request)


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₂₄H₄₆O₆
Storage Temperature: Ambient
MDL Number: MFCD00005366
CAS Number: 1338-41-6
EINECS: 215-664-9

Order Now

Specification Test Results

Assay 68% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR