D-(-)-Sorbitol ≥91%, powder NF

Supplier: SPECTRUM CHEMICAL MFG CORP

SO219-45KGBL SO219-2.5KG SO219-12KG SO219-500GM
700003-024EA 1673.6 USD
700003-024 95036-048 700003-022 95036-050
D-(-)-Sorbitol ≥91%, powder NF
D(-)-Sorbitol

Sorbitol, Powder, NF is used in the pharmaceutical industry as a sweetener or humectant. Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: HOH₂C(CH(OH))₄CH₂OH
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 94 - 98 °C
Density: 1.524 g/cm³ (20 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 91% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR