Safranine O

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: CI 50240

SA115-5KGBL SA115-100GM SA115-1KGBL SA115-25GM SA115-10GM
700010-212EA 3317.25 USD
700010-212 700010-204 700010-208 700010-210 700010-206
Safranine O
Safranine O

Safranine O, also known as Safranine, is used in cytology and histology as a biological stain.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Density: 1.447 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6
EINECS: 207-518-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR