Stearyl stearate ≥95%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Stearic acid stearyl ester, Octadecyl stearate, Octadecyl octadecanoate

S2017-100MG S2017-5GM
700005-040EA 211.65 USD
700005-040 700005-042
Stearyl stearate ≥95%
Stearyl stearate

Stearyl Stearate is used as a skin conditioning emollient and cosmetic viscosity control agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃₆H₇₂O₂
MW: 536.97 g/mol
Boiling Pt: ∼175 °C (1013 hPa)
Melting Pt: 64 °C
Flash Pt: >160 °C
Storage Temperature: Ambient
MDL Number: MFCD00056225
CAS Number: 2778-96-3
EINECS: 220-476-5

Order Now

Specification Test Results

Assay 95% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR