Sarcosine, Ultrapure
Supplier: Spectrum Chemical Mfg. Corp.
Sarcosine, Ultrapure, also known as N-methylglycine, is used in manufacturing biodegradable surfactants. The Ultrapure grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Ultrapure grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₃H₇NO₂ Boiling Pt: 185 °C (1013 hPa) Melting Pt: 63 °C Density: 1.301 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004279 CAS Number: 107-97-1 EINECS: 203-538-6 Merck Index: 12,08519 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...