Strontium hydroxide octahydrate ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

S1878-100GM S1878-2.5KG
700000-792EA 87.5 USD
700000-792 700000-796 700000-794
Strontium hydroxide octahydrate ≥98%
Strontium hydroxide octahydrate
Formula: H₂O₂Sr·8H₂O
MW: 265.76 g/mol
Melting Pt: 375 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150395
CAS Number: 1311-10-0
EINECS: 242-367-1
UN: 3262
ADR: 8,II

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR