Sulfuric acid ≥95% ACS

Supplier: Spectrum Chemical Mfg. Corp.
Danger

S1730-2.5LTP S1730-2.5LTG S1730-500MLG
700000-578EA 276.33 USD
700000-578 700000-576 700000-582
Sulfuric acid ≥95% ACS
Sulfuric acid

Sulfuric Acid, Reagent, ACS is an extremely corrosive acid that comes as yellowy slightly viscous liquid. It is soluble in water and is a diprotic acid. It has very strong corrosive, dehydrating and oxidizing properties as well as being hygroscopic. It uses range from detergent production, antifreeze, pharmaceuticals and fertilizers to cellophane, lubricants and production of explosives. As an ACS grade Reagent, Spectrum Chemical manufactured sulfuric acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness

Formula: H₂SO₄
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

Assay 95% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR