5-Sulfosalicylic acid dihydrate ≥99%, crystals ACS

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 2-Hydroxy-5-sulphobenzoic acid dihydrate

S1710-125GM S1710-2.5KG S1710-12KG S1710-500GM
700002-528EA 152.35 USD
700002-528 700002-532 700002-530 700002-536
5-Sulfosalicylic acid dihydrate ≥99%, crystals ACS
5-Sulphosalicylic acid dihydrate

5-Sulfosalicylic Acid, Dihydrate, Crystal, Reagent, ACS, also known simply as sulfosalicyclic acid, is used to determine urine protein content.


As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
MW: 254.22 g/mol
Melting Pt: 224 °C
Density: 1.695 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00149540
CAS Number: 5965-83-3
EINECS: 202-555-6
UN: 2585
ADR: 8,III
Merck Index: 13,09054
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69e00688850e885438e2fdd9a62f6d564ebf6ae8&vid=6a3840cf38254959a0062041e9a84fa6740b1288&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR