Strontium nitrate ≥99%, crystals ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

S1680-12KG S1680-125GM S1680-25GM S1680-2.5KGP S1680-500GM S1680-50KGBL
700001-630EA 2919.7 USD
700001-630 700001-628 700001-634 700001-632 700001-636 700001-638
Strontium nitrate ≥99%, crystals ACS for synthesis
Strontium nitrate

Strontium Nitrate, Crystal, Reagent, ACS is used as an oxidizer and colorant in pyrotechnics. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: Sr(NO₃)₂
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 570 °C
Density: 2.113 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011248
CAS Number: 10042-76-9
EINECS: 233-131-9
UN: 1507
ADR: 5.1,III
Merck Index: 13,08926

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR