Sudan orange G

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Solvent Orange 1, 4-(Phenylazo)resorcinol

S1634-25GM S1634-100GM S1634-5GM
700007-052EA 197.45 USD
700007-052 700007-050 700007-054
Sudan orange G
Sudan orange G

Sudan Orange G is useful for staining triglycerides in animal tissues (frozen sections).


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00002275
CAS Number: 2051-85-6
EINECS: 218-131-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR