Sodium sulphide hydrate 60%, fused flakes for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
di-Sodium sulphide hydrate
S1470-500GM
S1470-2.5KG
S1470-45KGBL
S1470-12KG
700001-408EA
154.45
USD
700001-408
700001-404
700001-406
700001-402
Sodium sulphide hydrate 60%, fused flakes for synthesis
Sodium sulphide hydrate
Sodium Sulfide, Fused Flakes, also known as disodium sulfide, is used as an oxygen scavenger agent in water treatment as well as a metals precipitant.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formula:
Na₂S·xH₂O Boiling Pt: ∼920 °C (1013 hPa) Melting Pt: 50 °C Density: ~ 1.43 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149183 CAS Number: 27610-45-3 EINECS: 215-211-5 UN: 1849 ADR: 8,II REACH: 01-2119513694-38 Merck Index: 12,08829 |
Specification Test Results
ASSAY (Na2S) | approx. 60% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...