di-Sodium succinate hexahydrate ≥98%, Reagent Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sodium succinate hexahydrate, Succinic acid disodium hexahydrate, 1,4-Butanedioic acid disodium salt hexahydrate, Sodium Succinate, Hexahydrate, Sodium succinate dibasic hexahydrate, Disodium succinate hexahydrate, Succinic acid disodium salt hexahydrate

S1449-100GM S1449-12KG S1449-500GM S1449-2.5KG
700005-960EA 95.61 USD
700005-960 700005-962 700005-966 700005-964
di-Sodium succinate hexahydrate ≥98%, Reagent Grade for synthesis
di-Sodium succinate hexahydrate

Sodium Succinate, Hexahydrate, Reagent, also known as succinic acid, is a diprotic and dicarboxylic acid. It is involved in an energy-yielding process known as the citric acid cycle. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: NaOOCCH₂CH₂COONa·6H₂O
Melting Pt: 300 °C
Density: 1.529 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149117
CAS Number: 6106-21-4
EINECS: 205-778-7
Merck Index: 13,08754

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR