Sodium stearate ≥90%, powder NF suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Stearate sodium, Stearic acid sodium salt

S1447-12KG S1447-2.5KG S1447-500GM
700005-032EA 1257.35 USD
700005-032 95035-946 95035-948
Sodium stearate ≥90%, powder NF suitable for use as excipient
Sodium stearate

Sodium Stearate, Powder, NF is used in the pharmaceutical industry as a surfactant to aid the solubility of hydrophobic compounds. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₁₈H₃₅NaO₂
MW: 306.46 g/mol
Melting Pt: 288
Density: 1.103 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036404
CAS Number: 822-16-2
EINECS: 212-490-5

Order Now

Specification Test Results

Assay 90% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR