Sodium silicate solution in aqueous solution 40 DEG Be

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

S1435-4LTGL S1435-200LTB S1435-20LT
700001-872EA 291.65 USD
700001-872 700001-868 700001-870
Sodium silicate solution in aqueous solution 40 DEG Be
Sodium silicate solutions

Sodium Silicate, 40 DEG Be Solution is typically used in cements, lumber processing and automobiles. It is also known as liquid glass and in pure composition is white or colorless. Some commercial samples, however, appear green or blue. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability

Formula: Na₂O(SiO₂)x·xH₂O
Density: 1.39
Storage Temperature: Ambient
MDL Number: MFCD00003492
CAS Number: 1344-09-8
EINECS: 215-687-4
UN: 3266
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR