Sodium hexadecyl sulfate
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Hexadecyl sulphuric acid salt
S1422-100GM
S1422-25GM
S1422-500GM
700015-734EA
300.5
USD
700015-734
700015-736
700015-738
Sodium hexadecyl sulfate
Sodium hexadecyl sulphate
Sodium n-Hexadecyl Sulfate is used as an anionic cleansing and emulsifying surfactant.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₆H₃₃NaO₄S MW: 344.49 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00047766 CAS Number: 1120-01-0 EINECS: 214-292-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...