Tetrasodium diphosphate, anhydrous ≥95%, powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Sodium pyrophosphate tetrabasic, Tetrasodium diphosphate, Sodium diphosphate, Sodium pyrophosphate, tetra-Sodium pyrophosphate

S1419-45KGBL S1419-12KG S1419-2.5KG S1419-500GM
89079-022EA 1579.45 USD
89079-022 700001-774 95035-916 95035-918
Tetrasodium diphosphate, anhydrous ≥95%, powder FCC
Tetrasodium diphosphate

Sodium Pyrophosphate, Anhydrous, Powder, FCC it is used as a palatability enhancer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Na₄O₇P₂
MW: 265.9 g/mol
Melting Pt: 988 °C
Density: 2.534 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003513
CAS Number: 7722-88-5
EINECS: 231-767-1
Merck Index: 13,09312

Order Now

Specification Test Results

Assay 95% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR