Disodium hydrogen phosphate heptahydrate ≥98%, granular ACS

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sodium phosphate dibasic heptahydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate

S1404-125GM S1404-2.5KG S1404-500GM
700001-654EA 109.27 USD
700001-654 700001-658 700001-660
Disodium hydrogen phosphate heptahydrate ≥98%, granular ACS
Disodium hydrogen phosphate heptahydrate

Sodium Phosphate, Dibasic, Heptahydrate, Granular, Reagent, ACS is used in foods and water treatment. In foods, it is used to adjust pH. Its presence prevents coagulation in the preparation of condensed milk. Similarly, it is used as an anti-caking additive in powdered products. In water treatment, It retards calcium scale formation. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Melting Pt: 48 °C
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS Number: 7782-85-6
EINECS: 231-448-7

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR