Stearic acid FCC

Supplier: Spectrum Chemical Mfg. Corp.

S1326-500GM S1326-2.5KG
95036-074EA 148.15 USD
95036-074 95036-072
Stearic acid FCC
Stearic acid

Stearic Acid, FCC is used as a lubricant, softening and release agent in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₈H₃₆O₂
Boiling Pt: 361 °C (1013 hPa)
Melting Pt: 67 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 196 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002752
CAS Number: 57-11-4
EINECS: 200-313-4
Merck Index: 13,08882
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b53ff33f46eb3558f286c08be23286aab54dce6c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR