Sodium iodate ≥99.5%, powder, Reagent Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

S1325-12KG S1325-2.5KG S1325-25GM S1325-100GM S1325-500GM S1325-50KGBL
700001-380EA 4941.25 USD
700001-380 700001-382 700001-384 700001-378 700001-386 700001-388
Sodium iodate ≥99.5%, powder, Reagent Grade for synthesis
Sodium iodate

Sodium Iodate, Powder, Reagent is an oxidizing agent that is the sodium salt of iodic acid. Sodium iodate is an oxidizing agent and known to start fires when in contact with combustible materials. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: INaO₃
MW: 197.89 g/mol
Melting Pt: 420…430 °C
Density: 3.56 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003533
CAS Number: 7681-55-2
EINECS: 231-672-5
UN: 1479
ADR: 5.1 II
Merck Index: 13,08704
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806173453dcec1d2f791367d269d662d3459149a&vid=16d5efbf45a7cc1cd6399272211e2455e47a2e4b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR