Sorbitan monostearate FCC

Supplier: Spectrum Chemical Mfg. Corp.

S1318-500GM S1318-2.5KG S1318-12KG
95036-018EA 199.45 USD
95036-018 95036-016 76014-866
Sorbitan monostearate FCC
Sorbitan monostearate

Sorbitan Monostearate, FCC is used as an emulsifier to keep oils and water mixed. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₂₄H₄₆O₆
Storage Temperature: Ambient
MDL Number: MFCD00005366
CAS Number: 1338-41-6
EINECS: 215-664-9

Order Now

Specification Test Results

Assay 27% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR