Sodium dehydracetate monohydrate ≥98% for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Sodium 3-Acetyl-4-hydroxy-6-methyl-2-oxo-4(2H)-pyranolate, Sodium Dehydroacetate monohydrate, Sodium dehydroacetate monohydrate, Dehydroacetic acid sodium salt monohydrate, Sodium 1-(3,4-dihydro-6-methyl-2,4-dioxo-2H-pyran-3-ylidene)ethonolate monohydrate, 3-Acetyl-4-hydroxy-6-methyl-2-pyrone sodium salt, Sodium Dehydroacetate Monohydrate, Dehydracetic acid sodium salt monohydrate

S1265-125GM S1265-2.5KG S1265-500GM
700008-424EA 118.05 USD
700008-424 700008-426 700008-430
Sodium dehydracetate monohydrate ≥98% for synthesis
Sodium dehydracetate monohydrate

Sodium Dehydroacetate is a water-soluble for cosmetics and toiletries (leave-on and rinse-off products).


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₈H₇NaO₄·1H₂O
Melting Pt: 280…285 °C
Storage Temperature: Ambient
MDL Number: MFCD00040583
CAS Number: 64039-28-7
EINECS: 224-580-1

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR