L(+)-Sodium lactate solution 60% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Sodium (S)-2-hydroxypropanoate, (S)-(+)-2-Hydroxypropanoic acid sodium salt, (S)-(+)-2-Hydroxypropionic acid sodium salt, Sodium-(S)-lactate, (S)-(+)-Lactic acid sodium salt, L(+)-Lactic acid sodium salt
Sodium Lactate Solution, 60 Percent, FCC is used in food products as a preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₃H₅NaO₃ MW: 112.06 g/mol Density: 1.270 Storage Temperature: Ambient |
MDL Number:
MFCD00066576 CAS Number: 867-56-1 EINECS: 212-762-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...