Sodium bromide ≥99%, granular ACS

Supplier: SPECTRUM CHEMICAL MFG CORP

S1195-500GM S1195-12KG S1195-2.5KG S1195-50KGBL S1195-125GM
700001-258EA 250.3 USD
700001-258 700001-254 700001-256 700001-260 700001-252
Sodium bromide ≥99%, granular ACS
Sodium bromide

Sodium Bromide, Granular, Reagent, ACS is used in photography in the preparation of silver bromide emulsion on photographic plates or films.


As a Reagent ACS grade reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: NaBr
MW: 102.89 g/mol
Boiling Pt: 1396 °C (1013 hPa)
Melting Pt: 753 °C
Density: 3.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6
EINECS: 231-599-9
Merck Index: 13,08666

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR