Sudan red G for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1-[(2-Methoxyphenyl)azo]-2-naphthol, Solvent Red 1, Oil Red 113

S1174-100GM S1174-25GM S1174-500GM
700015-630EA 345.95 USD
700015-630 700015-632 700015-634
Sudan red G for synthesis
Sudan red G

Solvent Red 1, also known as Sudan Red G, is a lysochrome azo dye used for coloring oils, fats, and waxes; it can also be used in hair dyes and temporary tattoos.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₇H₁₄N₂O₂
Melting Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00046377
CAS Number: 1229-55-6
EINECS: 214-968-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR